Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002857.1 | NP_001002857.1 | 14 | 1231 | ||
NM_001002858.2 | NP_001002858.1 | 13 | 1222 | |||
NM_001136015.2 | NP_001129487.1 | 14 | 1255 | |||
NM_004039.2 | NP_004030.1 | 13 | 1150 | |||
XM_011521477.2 | XP_011519779.1 | 12 | 1111 | |||
XM_017022090.1 | XP_016877579.1 | 16 | 4747 | |||
XM_017022091.1 | XP_016877580.1 | 16 | 4722 | |||
XM_017022092.1 | XP_016877581.1 | 13 | 1170 | |||
XM_017022093.1 | XP_016877582.1 | 12 | 1050 | |||
GenBank mRNA | AK092006.1 | - | 10 | 3310 | ||
AK124427.1 | - | 12 | 2119 | |||
AK222542.1 | - | 13 | 1149 | |||
AK297923.1 | - | 8 | 715 | |||
AK309356.1 | - | 1625 | ||||
BC001388.2 | - | 14 | 1219 | 3051155 | ||
BC001748.1 | - | 13 | 1305 | |||
BC009564.1 | - | 13 | 1132 | 3901641 | ||
BC013843.1 | - | 13 | 1143 | |||
BC015834.1 | - | 13 | 1154 | 4094952 | ||
BC016774.1 | - | 13 | 1156 | 4096060 | ||
BC021114.1 | - | 13 | 1132 | 5092810 | ||
BC023990.1 | - | 14 | 1235 | 3961419 | ||
BC052558.1 | - | 12 | 1106 | 6247665 | ||
BC052567.1 | - | 13 | 1137 | 6483607 | ||
BC066955.1 | - | 13 | 1150 | |||
BC068065.1 | - | 13 | 1130 | 6265952 | ||
BC093056.1 | - | 14 | 1255 | 30403089 | ||
BX640598.1 | - | 13 | 1221 | |||
D00017.1 | - | 13 | 1144 | |||
FJ224302.1 | - | 14 | 1235 |