Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H2AE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021052.2 | NP_066390.1 | 1 | 175 | ||
GenBank mRNA | AK311824.1 | - | 1 | 174 | ||
BC093836.1 | - | 1 | 146 | 7939681 | ||
BC093862.1 | - | 1 | 146 | 7939707 |