Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007067.1 | NP_001007068.1 | 9 | 1913 | ||
NM_001007068.1 | NP_001007069.1 | 9 | 1898 | |||
NM_001007069.1 | NP_001007070.1 | 9 | 1913 | |||
NM_001007070.1 | NP_001007071.1 | 9 | 1910 | |||
NM_005625.3 | NP_005616.2 | 9 | 1916 | |||
XM_011517571.2 | XP_011515873.2 | 9 | 3285 | |||
XM_017013727.1 | XP_016869216.1 | 9 | 3288 | |||
XM_017013728.1 | XP_016869217.1 | 9 | 3285 | |||
GenBank mRNA | AF000652.1 | - | 9 | 1913 | ||
AF006636.1 | - | 9 | 1840 | |||
AK128645.1 | - | 10 | 3338 | |||
BC013254.2 | - | 9 | 1800 | |||
BG111210.1 | - | 1 | 376 | |||
BU160865.1 | - | 1 | 657 | |||
U83463.1 | - | 8 | 1790 |