Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS1 SIRNA Assays ›
Gene Symbol:
SORBS1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location:
Chr.10: 95311773 - 95561420 on Build GRCh38
UniGene ID:
Hs.38621
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
288596
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034954.1
NP_001030126.1
9
1013
NM_001034955.1
NP_001030127.1
9
1013
NM_001034956.1
NP_001030128.1
9
806
NM_001034957.1
NP_001030129.1
6
521
NM_001290294.1
NP_001277223.1
11
1200
NM_001290295.1
NP_001277224.1
9
735
NM_001290296.1
NP_001277225.1
8
986
NM_001290297.1
NP_001277226.1
7
548
NM_001290298.1
NP_001277227.1
7
617
NM_006434.2
NP_006425.2
6
521
NM_015385.3
NP_056200.1
10
1104
NM_024991.2
NP_079267.1
10
831
XM_006717589.1
XP_006717652.1
10
1126
XM_006717593.1
XP_006717656.1
8
686
XM_011539140.1
XP_011537442.1
12
1870
XM_011539150.1
XP_011537452.1
12
1870
XM_011539155.1
XP_011537457.1
10
1772
XM_011539167.1
XP_011537469.1
9
1055
XM_017015500.1
XP_016870989.1
12
1870
XM_017015501.1
XP_016870990.1
12
1870
XM_017015502.1
XP_016870991.1
12
1870
XM_017015503.1
XP_016870992.1
12
1870
XM_017015504.1
XP_016870993.1
10
784
XM_017015505.1
XP_016870994.1
12
1870
XM_017015506.1
XP_016870995.1
10
784
XM_017015507.1
XP_016870996.1
12
1870
XM_017015508.1
XP_016870997.1
10
784
XM_017015509.1
XP_016870998.1
9
713
XM_017015510.1
XP_016870999.1
12
1870
XM_017015511.1
XP_016871000.1
10
784
XM_017015512.1
XP_016871001.1
8
686
XM_017015513.1
XP_016871002.1
8
686
XM_017015514.1
XP_016871003.1
8
686
XM_017015515.1
XP_016871004.1
11
1153
XM_017015516.1
XP_016871005.1
10
1126
XM_017015517.1
XP_016871006.1
11
1153
XM_017015518.1
XP_016871007.1
10
1082
XM_017015519.1
XP_016871008.1
9
1055
XM_017015520.1
XP_016871009.1
11
1153
XM_017015521.1
XP_016871010.1
10
1082
XM_017015522.1
XP_016871011.1
10
1126
XM_017015523.1
XP_016871012.1
10
1082
XM_017015524.1
XP_016871013.1
9
1055
XM_017015525.1
XP_016871014.1
9
713
XM_017015526.1
XP_016871015.1
10
784
XM_017015527.1
XP_016871016.1
9
713
XM_017015528.1
XP_016871017.1
9
757
XM_017015529.1
XP_016871018.1
10
784
XM_017015530.1
XP_016871019.1
10
784
XM_017015531.1
XP_016871020.1
9
713
XM_017015532.1
XP_016871021.1
9
757
XM_017015533.1
XP_016871022.1
8
686
XM_017015534.1
XP_016871023.1
10
784
XM_017015535.1
XP_016871024.1
9
713
XM_017015536.1
XP_016871025.1
9
713
XM_017015537.1
XP_016871026.1
9
757
XM_017015538.1
XP_016871027.1
9
757
XM_017015539.1
XP_016871028.1
8
686
XM_017015540.1
XP_016871029.1
8
686
GenBank mRNA
AB037717.1
-
10
763
AF136380.1
-
5
476
AF136381.1
-
7
941
AF330623.1
-
6
503
AF356525.1
-
9
1024
AF356526.1
-
9
1024
AF356527.1
-
9
817
AJ489942.1
-
6
572
AK300407.1
-
9
688
AK304753.1
-
10
1747
AL117472.1
-
10
1062
AM260536.1
-
11
1158
BC042612.1
-
8
1128
BC152463.1
-
10
763
8862599
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell-matrix adhesion
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
actin binding
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
cytoskeletal protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0