Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS1 SIRNA Assays ›
Gene Symbol:
SORBS1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location:
Chr.10: 95311773 - 95561420 on Build GRCh38
UniGene ID:
Hs.38621
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
288597
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034954.1
NP_001030126.1
31
6224
NM_001034955.1
NP_001030127.1
31
5801
NM_001034956.1
NP_001030128.1
28
5063
NM_001034957.1
NP_001030129.1
22
4568
NM_001290294.1
NP_001277223.1
32
6333
NM_001290295.1
NP_001277224.1
25
4782
NM_001290296.1
NP_001277225.1
28
5360
NM_001290297.1
NP_001277226.1
25
4781
NM_001290298.1
NP_001277227.1
26
4886
NM_006434.2
NP_006425.2
21
4400
NM_015385.3
NP_056200.1
25
4983
NM_024991.2
NP_079267.1
26
4878
XM_006717589.1
XP_006717652.1
30
5500
XM_006717593.1
XP_006717656.1
24
4733
XM_011539140.1
XP_011537442.1
34
7078
XM_011539150.1
XP_011537452.1
33
7003
XM_011539155.1
XP_011537457.1
28
6653
XM_011539167.1
XP_011537469.1
27
5936
XM_017015500.1
XP_016870989.1
29
6583
XM_017015501.1
XP_016870990.1
31
6169
XM_017015502.1
XP_016870991.1
30
6076
XM_017015503.1
XP_016870992.1
29
6001
XM_017015504.1
XP_016870993.1
32
5992
XM_017015505.1
XP_016870994.1
28
5917
XM_017015506.1
XP_016870995.1
31
5917
XM_017015507.1
XP_016870996.1
29
5908
XM_017015508.1
XP_016870997.1
30
5824
XM_017015509.1
XP_016870998.1
30
5846
XM_017015510.1
XP_016870999.1
27
5749
XM_017015511.1
XP_016871000.1
29
5740
XM_017015512.1
XP_016871001.1
27
5651
XM_017015513.1
XP_016871002.1
27
5642
XM_017015514.1
XP_016871003.1
27
5558
XM_017015515.1
XP_016871004.1
29
5359
XM_017015516.1
XP_016871005.1
28
5332
XM_017015517.1
XP_016871006.1
28
5275
XM_017015518.1
XP_016871007.1
28
5288
XM_017015519.1
XP_016871008.1
27
5261
XM_017015520.1
XP_016871009.1
27
5200
XM_017015521.1
XP_016871010.1
27
5213
XM_017015522.1
XP_016871011.1
26
5173
XM_017015523.1
XP_016871012.1
26
5129
XM_017015524.1
XP_016871013.1
25
5102
XM_017015525.1
XP_016871014.1
29
5087
XM_017015526.1
XP_016871015.1
28
4990
XM_017015527.1
XP_016871016.1
28
5012
XM_017015528.1
XP_016871017.1
27
4963
XM_017015529.1
XP_016871018.1
27
4915
XM_017015530.1
XP_016871019.1
27
4906
XM_017015531.1
XP_016871020.1
27
4919
XM_017015532.1
XP_016871021.1
26
4888
XM_017015533.1
XP_016871022.1
26
4892
XM_017015534.1
XP_016871023.1
27
4822
XM_017015535.1
XP_016871024.1
26
4844
XM_017015536.1
XP_016871025.1
26
4835
XM_017015537.1
XP_016871026.1
25
4804
XM_017015538.1
XP_016871027.1
26
4795
XM_017015539.1
XP_016871028.1
25
4817
XM_017015540.1
XP_016871029.1
25
4808
GenBank mRNA
AB037717.1
-
26
4811
AL117472.1
-
25
4941
AL122082.1
-
1
1641
AL832353.1
-
6
3406
AM260536.1
-
32
6292
BC152463.1
-
26
4811
8862599
BX640814.1
-
15
4873
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell-matrix adhesion
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
actin binding
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
cytoskeletal protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline