Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007231.2 | NP_001007232.2 | 11 | 2569 | ||
NM_001166276.1 | NP_001159748.1 | 10 | 2509 | |||
NM_001166277.1 | NP_001159749.1 | 10 | 2449 | |||
NM_014882.2 | NP_055697.1 | 10 | 2506 | |||
XM_005264675.2 | XP_005264732.1 | 11 | 2273 | |||
XM_005264676.2 | XP_005264733.1 | 10 | 2157 | |||
XM_011533206.2 | XP_011531508.1 | 11 | 2317 | |||
XM_011533207.2 | XP_011531509.1 | 11 | 2283 | |||
XM_011533208.2 | XP_011531510.1 | 12 | 2334 | |||
XM_011533209.2 | XP_011531511.1 | 8 | 1874 | |||
XM_011533210.2 | XP_011531512.1 | 6 | 1401 | |||
XM_017005423.1 | XP_016860912.1 | 11 | 2177 | |||
XM_017005424.1 | XP_016860913.1 | 11 | 2174 | |||
XM_017005425.1 | XP_016860914.1 | 10 | 2060 | |||
XM_017005426.1 | XP_016860915.1 | 8 | 2511 | |||
XM_017005427.1 | XP_016860916.1 | 8 | 2508 | |||
GenBank mRNA | AK290396.1 | - | 11 | 2247 | ||
AK297157.1 | - | 11 | 2309 | |||
AK298669.1 | - | 11 | 2122 | |||
D29642.1 | - | 10 | 2319 | |||
EU794656.1 | - | 10 | 2506 |