Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040118.2 | NP_001035207.1 | 27 | 3859 | ||
NM_001135190.1 | NP_001128662.1 | 24 | 3487 | |||
NM_015242.4 | NP_056057.2 | 25 | 3670 | |||
GenBank mRNA | AB018325.2 | - | 25 | 3647 | ||
AF411983.1 | - | 21 | 2728 | |||
AJ621557.1 | - | 25 | 3670 | |||
AK160383.1 | - | 23 | 4337 | |||
AY049732.1 | - | 22 | 3507 | |||
AY553630.1 | - | 27 | 3874 | |||
BC008315.1 | - | 9 | 1140 | |||
BC056401.1 | - | 13 | 1513 | |||
BC140792.1 | - | 24 | 3157 | 9021660 | ||
BC144562.1 | - | 25 | 3340 | 9053090 |