Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002211.3 | NP_002202.2 | 7, 8 | 1161 | ||
NM_033668.2 | NP_391988.1 | 6, 7 | 940 | |||
NM_133376.2 | NP_596867.1 | 7, 8 | 1076 | |||
GenBank mRNA | AK291697.1 | - | 7, 8 | 1013 | ||
BC020057.1 | - | 7, 8 | 1005 | 3924411 | ||
BC113901.1 | - | 1, 2 | 162 | |||
BX537407.1 | - | 7, 8 | 1015 | |||
CX868374.1 | - | 3, 4 | 418 | |||
CX871886.1 | - | 3, 4 | 569 |