Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321309.1 | NP_001308238.1 | 33 | 7158 | ||
NM_053025.3 | NP_444253.3 | 34 | 7366 | |||
NM_053026.3 | NP_444254.3 | 33 | 7159 | |||
NM_053027.3 | NP_444255.3 | 33 | 7213 | |||
NM_053028.3 | NP_444256.3 | 32 | 7006 | |||
XM_017006472.1 | XP_016861961.1 | 4 | 1976 | |||
XM_017006473.1 | XP_016861962.1 | 4 | 1925 | |||
GenBank mRNA | AF172084.1 | - | 1 | 1231 | ||
AK025953.1 | - | 1 | 1138 | |||
AK127437.1 | - | 1 | 2887 | |||
BC015838.1 | - | 1 | 489 | |||
BC064420.1 | - | 3 | 2038 | |||
BC113456.1 | - | 3 | 1847 | |||
BC113458.1 | - | 3 | 1842 | |||
CR749364.1 | - | 3 | 6825 |