Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000268.3 | NP_000259.1 | 1 | 480 | ||
NM_016418.5 | NP_057502.2 | 1 | 480 | |||
NM_181825.2 | NP_861546.1 | 1 | 480 | |||
NM_181828.2 | NP_861966.1 | 1 | 480 | |||
NM_181829.2 | NP_861967.1 | 1 | 480 | |||
NM_181830.2 | NP_861968.1 | 1 | 480 | |||
NM_181831.2 | NP_861969.1 | 1 | 480 | |||
NM_181832.2 | NP_861970.1 | 1 | 480 | |||
NM_181833.2 | NP_861971.1 | 1 | 480 | |||
GenBank mRNA | AF122827.1 | - | 1 | 87 | ||
AF122828.1 | - | 1 | 87 | |||
AF123570.1 | - | 1 | 87 | |||
AF369657.1 | - | 1 | 462 | |||
AF369658.1 | - | 1 | 462 | |||
AF369661.1 | - | 1 | 37 | |||
AF369662.1 | - | 1 | 37 | |||
AF369663.1 | - | 1 | 37 | |||
AF369664.1 | - | 1 | 37 | |||
AF369665.1 | - | 1 | 37 | |||
AF369666.1 | - | 1 | 37 | |||
AF369667.1 | - | 1 | 37 | |||
AF369668.1 | - | 1 | 37 | |||
AF369701.1 | - | 1 | 403 | |||
AK297116.1 | - | 1 | 419 | |||
BC003112.1 | - | 1 | 309 | 3537045 | ||
BC020257.1 | - | 1 | 375 | 4871980 | ||
CF138178.1 | - | 1 | 261 | |||
CN419318.1 | - | 1 | 480 | |||
CR456530.1 | - | 1 | 99 | |||
EF517524.1 | - | 1 | 37 | |||
L11353.1 | - | 1 | 256 | |||
Z22664.1 | - | 1 | 211 |