Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300981.1 | NP_001287910.1 | 6 | 1004 | ||
NM_021130.4 | NP_066953.1 | 5 | 847 | |||
GenBank mRNA | AK130101.1 | - | 5 | 801 | ||
AK290085.1 | - | 5 | 803 | |||
AK290851.1 | - | 5 | 805 | |||
AK297340.1 | - | 6 | 694 | |||
BM909285.1 | - | 524 | ||||
BX347265.1 | - | 1 | 72 | |||
BX408703.2 | - | 1 | 72 |