Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135050.1 | NP_001128522.1 | 7 | 995 | ||
NM_020789.3 | NP_065840.2 | 7 | 995 | |||
XM_017001884.1 | XP_016857373.1 | 7 | 1060 | |||
GenBank mRNA | AB037776.1 | - | 7 | 995 | ||
AY358128.1 | - | 7 | 1013 | |||
BC030141.2 | - | 7 | 995 | 4762143 |