Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020886.3 | NP_065937.1 | 3 | 269 | ||
XM_005271630.2 | XP_005271687.1 | 3 | 265 | |||
XM_005271631.2 | XP_005271688.1 | 3 | 265 | |||
XM_005271632.2 | XP_005271689.1 | 4 | 595 | |||
XM_005271633.4 | XP_005271690.1 | 4 | 323 | |||
XM_005271636.2 | XP_005271693.1 | 3 | 265 | |||
XM_005271637.2 | XP_005271694.1 | 3 | 265 | |||
XM_005271638.2 | XP_005271695.1 | 3 | 265 | |||
XM_005271639.2 | XP_005271696.1 | 3 | 265 | |||
XM_011542936.2 | XP_011541238.1 | 2 | 184 | |||
XM_017018056.1 | XP_016873545.1 | 2 | 184 | |||
XM_017018057.1 | XP_016873546.1 | 4 | 595 | |||
XM_017018058.1 | XP_016873547.1 | 2 | 201 | |||
XM_017018059.1 | XP_016873548.1 | 2 | 184 | |||
XM_017018063.1 | XP_016873552.1 | 3 | 219 | |||
XM_017018064.1 | XP_016873553.1 | 2 | 140 | |||
XM_017018065.1 | XP_016873554.1 | 3 | 233 | |||
XM_017018066.1 | XP_016873555.1 | 4 | 595 | |||
GenBank mRNA | AF266283.1 | - | 3 | 206 | ||
AK097623.1 | - | 3 | 265 | |||
BC065928.1 | - | 3 | 227 | 6140725 | ||
DB061316.1 | - | 3 | 269 |