Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003345.4 | NP_003336.1 | 3 | 270 | ||
NM_194259.2 | NP_919235.1 | 4 | 527 | |||
NM_194260.2 | NP_919236.1 | 3 | 390 | |||
NM_194261.2 | NP_919237.1 | 3 | 242 | |||
XM_005255540.2 | XP_005255597.1 | 3 | 261 | |||
XM_005255541.4 | XP_005255598.1 | 3 | 667 | |||
XM_011522644.2 | XP_011520946.1 | 4 | 507 | |||
XM_011522645.1 | XP_011520947.1 | 3 | 372 | |||
XM_017023640.1 | XP_016879129.1 | 3 | 667 | |||
GenBank mRNA | AB208988.1 | - | 3 | 351 | ||
AJ002385.1 | - | 3 | 190 | |||
AK093616.1 | - | 3 | 201 | |||
AK291118.1 | - | 3 | 261 | |||
BC000427.2 | - | 3 | 174 | 2819764 | ||
BC004429.1 | - | 3 | 174 | 2819764 | ||
BC051289.1 | - | 3 | 204 | |||
BE262845.1 | - | 4 | 509 | |||
BF315618.1 | - | 4 | 486 | |||
BI523501.1 | - | 3 | 380 | |||
BT006932.1 | - | 2 | 112 | |||
BT007010.1 | - | 2 | 112 | |||
BU543455.1 | - | 3 | 262 | |||
CD249028.1 | - | 3 | 211 | |||
CX788607.1 | - | 3 | 346 | |||
D45050.1 | - | 3 | 229 | |||
U29092.1 | - | 2 | 136 | |||
U31882.1 | - | 3 | 205 | |||
U31933.1 | - | 3 | 230 | |||
U38785.1 | - | 3 | 204 | |||
U45328.1 | - | 3 | 235 | |||
U66818.1 | - | 3 | 199 | |||
U66867.1 | - | 3 | 918 | |||
X96427.1 | - | 3 | 235 |