Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LLGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015002.1 | NP_001015002.1 | 6 | 618 | ||
NM_001031803.1 | NP_001026973.1 | 6 | 618 | |||
NM_004524.2 | NP_004515.2 | 6 | 618 | |||
XM_011524801.1 | XP_011523103.1 | 6 | 601 | |||
XM_011524802.1 | XP_011523104.1 | 6 | 589 | |||
XM_011524807.2 | XP_011523109.1 | 6 | 706 | |||
XM_017024625.1 | XP_016880114.1 | 6 | 589 | |||
XM_017024626.1 | XP_016880115.1 | 6 | 589 | |||
XM_017024627.1 | XP_016880116.1 | 6 | 589 | |||
XM_017024628.1 | XP_016880117.1 | 6 | 589 | |||
XM_017024629.1 | XP_016880118.1 | 6 | 589 | |||
XM_017024630.1 | XP_016880119.1 | 6 | 589 | |||
XM_017024631.1 | XP_016880120.1 | 6 | 589 | |||
GenBank mRNA | AK126687.1 | - | 3 | 641 | ||
AK315126.1 | - | 6 | 568 | |||
BC006503.2 | - | 7 | 676 | |||
BC010879.1 | - | 6 | 619 | |||
BC064994.1 | - | 6 | 591 | 5575350 | ||
X87342.1 | - | 6 | 594 |