Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMY2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000699.3 | NP_000690.1 | 4 | 639 | ||
GenBank mRNA | AK223463.1 | - | 4 | 554 | ||
BC007060.1 | - | 4 | 594 | 3949799 | ||
BC146997.1 | - | 4 | 639 | 9056925 | ||
M28443.1 | - | 4 | 543 |