Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000771.3 | NP_000762.2 | 9 | 1604 | ||
XM_017015758.1 | XP_016871247.1 | 10 | 2024 | |||
GenBank mRNA | AK289420.1 | - | 9 | 1604 | ||
BC125054.1 | - | 9 | 1579 | 40116637 | ||
D00173.1 | - | 9 | 1571 | |||
M21940.1 | - | 7 | 1256 | |||
M61855.1 | - | 9 | 1591 | |||
M61857.1 | - | 9 | 1589 | |||
S46963.1 | - | 9 | 1542 |