Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000795.3 | NP_000786.1 | 8 | 2453 | ||
NM_016574.3 | NP_057658.2 | 7 | 2366 | |||
XM_017017296.1 | XP_016872785.1 | 8 | 2435 | |||
GenBank mRNA | AB209832.1 | - | 7 | 2248 | ||
BC021195.2 | - | 8 | 2313 | 3939741 | ||
M29066.1 | - | 7 | 2250 | |||
S69899.1 | - | 7 | 2227 | |||
X51362.1 | - | 8 | 2367 |
Set Membership: |
G-protein coupled receptor |