Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002911.3 | NP_001002911.1 | 2 | 538 | ||
NM_001318483.1 | NP_001305412.1 | 3 | 637 | |||
GenBank mRNA | AB196529.1 | - | 2 | 376 | ||
AK291384.1 | - | 2 | 677 | |||
AY635179.1 | - | 2 | 467 | |||
BC137237.1 | - | 2 | 527 | 9021234 | ||
BC137238.1 | - | 2 | 527 | 9021235 | ||
BC144547.1 | - | 3 | 626 | |||
DN693347.1 | - | 2 | 538 | |||
KR081941.1 | - | 2 | 376 |