Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000535.6 | NP_000526.2 | 5, 6 | 632 | ||
NM_001322003.1 | NP_001308932.1 | 5, 6 | 627 | |||
NM_001322004.1 | NP_001308933.1 | 4, 5 | 492 | |||
NM_001322005.1 | NP_001308934.1 | 5, 6 | 744 | |||
NM_001322006.1 | NP_001308935.1 | 5, 6 | 632 | |||
NM_001322007.1 | NP_001308936.1 | 4, 5 | 529 | |||
NM_001322008.1 | NP_001308937.1 | 3, 4 | 389 | |||
NM_001322009.1 | NP_001308938.1 | 5, 6 | 739 | |||
NM_001322010.1 | NP_001308939.1 | 4, 5 | 492 | |||
NM_001322014.1 | NP_001308943.1 | 5, 6 | 632 | |||
NM_001322015.1 | NP_001308944.1 | 5, 6 | 802 | |||
NR_136154.1 | 5, 6 | 632 | ||||
GenBank mRNA | AB103082.1 | - | 5, 6 | 545 | ||
AB103083.1 | - | 5, 6 | 545 | |||
AB103086.1 | - | 2, 3 | 227 | |||
AK294661.1 | - | 4, 5 | 482 | |||
AK312390.1 | - | 5, 6 | 581 | |||
BC008400.1 | - | 5, 6 | 590 | |||
BC031832.1 | - | 5, 6 | 596 | |||
BC093921.1 | - | 5, 6 | 567 | 7939766 | ||
BP251389.1 | - | 1, 2 | 102 | |||
CR981250.1 | - | 5, 6 | 533 | |||
U14658.1 | - | 5, 6 | 547 |