Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEFTY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020997.3 | NP_066277.1 | 4 | 1361 | ||
GenBank mRNA | AF081512.1 | - | 4 | 1351 | ||
AK129605.1 | - | 4 | 1064 | |||
AY358873.1 | - | 4 | 1348 | |||
BC027883.1 | - | 4 | 1348 | 5221120 |