Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082.4 | NP_001073.3 | 13 | 1819 | ||
GenBank mRNA | AK290790.1 | - | 13 | 1773 | ||
AK296200.1 | - | 11 | 1666 | |||
D26480.1 | - | 13 | 1764 | |||
U02388.2 | - | 13 | 1782 |
Gene Symbol: CYP4F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000896.2 | NP_000887.2 | 13 | 1773 | ||
NM_001199208.1 | NP_001186137.1 | 13 | 1773 | |||
NM_001199209.1 | NP_001186138.1 | 13 | 1757 | |||
XM_005259911.4 | XP_005259968.1 | 11 | 1701 | |||
XM_011528014.2 | XP_011526316.1 | 11 | 1550 | |||
XM_017026814.1 | XP_016882303.1 | 12 | 1851 | |||
GenBank mRNA | AF054821.1 | - | 12 | 1754 | ||
AK296761.1 | - | 13 | 1692 | |||
AK298675.1 | - | 13 | 1753 | |||
AK303477.1 | - | 7 | 1116 | |||
AK304200.1 | - | 13 | 1770 | |||
BC136299.1 | - | 13 | 1765 | 9020286 |