Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001120.4 | NP_001111.3 | 12 | 1893 | ||
NM_001146069.1 | NP_001139541.1 | 13 | 1691 | |||
XM_005247927.3 | XP_005247984.1 | 12 | 1793 | |||
GenBank mRNA | BC001502.2 | - | 13 | 1617 | 2988084 | |
BC014979.1 | - | 13 | 1620 | 4878602 | ||
L11669.1 | - | 12 | 1699 |