Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001402.5 | NP_001393.1 | 8 | 1800 | ||
GenBank mRNA | AJ420488.1 | - | 7 | 1760 | ||
AK304214.1 | - | 9 | 1760 |