Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001632.4 | NP_001623.3 | 11 | 2950 | ||
GenBank mRNA | AF217992.1 | - | 1 | 1107 | ||
BC068501.1 | - | 11 | 2733 | 30349481 | ||
BC094743.1 | - | 11 | 2813 | 30330997 | ||
M14170.1 | - | 11 | 2694 |