Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030047.1 | NP_001025218.1 | 2 | 189 | ||
NM_001030048.1 | NP_001025219.1 | 2 | 189 | |||
NM_001648.2 | NP_001639.1 | 2 | 189 | |||
GenBank mRNA | AF335477.1 | - | 2 | 612 | ||
AF335478.1 | - | 1 | 1427 | |||
AJ310937.1 | - | 2 | 185 | |||
AJ310938.1 | - | 2 | 185 | |||
AJ459782.1 | - | 2 | 169 | |||
AJ459783.1 | - | 2 | 169 | |||
AJ512346.1 | - | 2 | 169 | |||
AK312933.1 | - | 2 | 183 | |||
AX772834.1 | - | 2 | 189 | |||
BC005307.1 | - | 2 | 189 | 3950475 | ||
BC050595.2 | - | 2 | 212 | 6199928 | ||
BC056665.1 | - | 2 | 194 | 4296876 | ||
BF679511.1 | - | 2 | 190 | |||
BQ932072.1 | - | 2 | 155 | |||
BT019862.1 | - | 2 | 148 | |||
M21895.1 | - | 2 | 137 | |||
M21896.1 | - | 2 | 137 | |||
M21897.1 | - | 2 | 137 | |||
M26663.1 | - | 2 | 172 | |||
S75755.1 | - | 2 | 189 | |||
U17040.1 | - | 2 | 189 | |||
X05332.1 | - | 2 | 191 | |||
X07730.1 | - | 1 | 453 |