Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001651.3 | NP_001642.1 | 1 | 592 | ||
XM_005268838.2 | XP_005268895.1 | 1 | 233 | |||
GenBank mRNA | BC032946.1 | - | 1 | 142 | 5269384 | |
BQ083334.1 | - | 1 | 222 | |||
CR541989.1 | - | 1 | 70 | |||
CR542022.1 | - | 1 | 70 |