Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001823.4 | NP_001814.2 | 8 | 1434 | ||
XM_017020951.1 | XP_016876440.1 | 7 | 1480 | |||
GenBank mRNA | AK290101.1 | - | 8 | 1405 | ||
AK298198.1 | - | 8 | 1303 | |||
BC001190.1 | - | 8 | 1393 | 3354679 | ||
BC004914.2 | - | 8 | 1385 | 3537262 | ||
BC008323.1 | - | 8 | 1391 | 3358085 | ||
BC010002.2 | - | 8 | 1400 | 4135106 | ||
BC019259.1 | - | 8 | 1389 | 3503057 | ||
BC019281.1 | - | 8 | 1399 | 2823530 | ||
EU794629.1 | - | 8 | 1389 | |||
L47647.1 | - | 8 | 1371 | |||
M16364.1 | - | 7 | 1338 | |||
M16451.1 | - | 7 | 1352 |