Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318915.1 | NP_001305844.1 | 5 | 1254 | ||
NM_002338.4 | NP_002329.2 | 5 | 1254 | |||
XM_011512840.2 | XP_011511142.1 | 5 | 1258 | |||
XM_017006383.1 | XP_016861872.1 | 5 | 1258 | |||
XM_017006384.1 | XP_016861873.1 | 4 | 476 | |||
GenBank mRNA | AK299851.1 | - | 1069 | |||
AK315286.1 | - | 5 | 1193 | |||
BC033803.1 | - | 5 | 1247 | 5188760 | ||
U41901.1 | - | 5 | 747 |