Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184986.1 | NP_001171915.1 | 6 | 1185 | ||
NM_001184987.1 | NP_001171916.1 | 5 | 653 | |||
NM_001184988.1 | NP_001171917.1 | 6 | 689 | |||
NM_001184989.1 | NP_001171918.1 | 6 | 712 | |||
NM_001184990.1 | NP_001171919.1 | 5 | 649 | |||
NM_001184991.1 | NP_001171920.1 | 5 | 1126 | |||
NM_002494.3 | NP_002485.1 | 5 | 788 | |||
GenBank mRNA | AF047184.1 | - | 4 | 383 | ||
AF047435.1 | - | 4 | 363 | |||
AK291147.1 | - | 4 | 393 | |||
BC107682.1 | - | 5 | 758 | 5557684 | ||
CN347371.1 | - | 5 | 554 |