Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021021.3 | NP_066301.1 | 1, 2 | 789 | ||
XM_011517239.1 | XP_011515541.1 | 1, 2 | 766 | |||
GenBank mRNA | AF028828.1 | - | 1, 2 | 283 | ||
AK026095.1 | - | 3, 4 | 932 | |||
AK292655.1 | - | 1, 2 | 863 | |||
BC098573.1 | - | 1, 2 | 749 | 30925462 |