Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002928.3 | NP_002919.3 | 5 | 1755 | ||
GenBank mRNA | U70426.1 | - | 5 | 1693 | ||
U94829.1 | - | 5 | 1601 |