Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303252.1 | NP_001290181.1 | 11 | 1516 | ||
NM_002969.4 | NP_002960.2 | 12 | 1546 | |||
GenBank mRNA | BC015741.1 | - | 12 | 1447 | 4907996 | |
BX640648.1 | - | 10 | 1153 | |||
X79483.1 | - | 12 | 1329 | |||
Y10487.1 | - | 12 | 1502 |