Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304464.1 | NP_001291393.1 | 5 | 1500 | ||
NM_003094.3 | NP_003085.1 | 5 | 1456 | |||
NR_130746.1 | 4 | 1459 | ||||
GenBank mRNA | BC072433.1 | - | 5 | 1404 | ||
BC090951.1 | - | 5 | 1429 | 5204266 |