Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256510.1 | NP_001243439.1 | 6 | 820 | ||
NM_001256511.1 | NP_001243440.1 | 6 | 701 | |||
NM_001256512.1 | NP_001243441.1 | 6 | 683 | |||
NM_001256513.1 | NP_001243442.1 | 6 | 611 | |||
NM_003143.2 | NP_003134.1 | 6 | 518 | |||
NR_046269.1 | 7 | 568 | ||||
XM_005250048.4 | XP_005250105.1 | 6 | 530 | |||
XM_005250049.4 | XP_005250106.1 | 6 | 569 | |||
XM_005250050.4 | XP_005250107.1 | 6 | 620 | |||
XM_005250051.4 | XP_005250108.1 | 6 | 646 | |||
GenBank mRNA | AF277319.1 | - | 6 | 464 | ||
AK294393.1 | - | 6 | 474 | |||
AK313033.1 | - | 6 | 531 | |||
BC000895.1 | - | 6 | 820 | 3464282 | ||
BC093054.1 | - | 6 | 480 | 30722138 | ||
BF131408.1 | - | 7 | 499 | |||
BP308841.1 | - | 5 | 387 | |||
BQ959702.1 | - | 6 | 583 | |||
M94556.1 | - | 6 | 463 |