Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032283.2 | NP_001027454.1 | 2 | 611 | ||
NM_001032284.2 | NP_001027455.1 | 2 | 611 | |||
NM_001307975.1 | NP_001294904.1 | 2 | 611 | |||
NM_003276.2 | NP_003267.1 | 2 | 611 | |||
XM_005269132.3 | XP_005269189.1 | 2 | 602 | |||
XM_017019914.1 | XP_016875403.1 | 2 | 303 | |||
GenBank mRNA | AB209297.1 | - | 2 | 548 | ||
AF070631.1 | - | 2 | 552 | |||
AK315221.1 | - | 2 | 541 | |||
BC053675.1 | - | 2 | 557 | 6170639 | ||
EF028063.1 | - | 2 | 332 | |||
S76736.1 | - | 2 | 516 | |||
U09086.1 | - | 2 | 520 | |||
U09087.1 | - | 2 | 553 | |||
U09088.1 | - | 2 | 553 |