Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003302.2 | NP_003293.2 | 9 | 1684 | ||
GenBank mRNA | AF000974.1 | - | 9 | 1673 | ||
AF025437.1 | - | 6 | 857 | |||
AJ001902.1 | - | 9 | 1586 | |||
AK056773.1 | - | 8 | 1688 | |||
AK291906.1 | - | 9 | 1621 | |||
BC002680.2 | - | 9 | 1577 | 3610413 | ||
BC004249.1 | - | 9 | 1589 | 3621877 | ||
BC004999.1 | - | 9 | 1583 | 2958142 | ||
BC021540.1 | - | 9 | 1589 | 3622017 | ||
BC028985.1 | - | 9 | 1596 | 5122169 | ||
CR749505.1 | - | 8 | 2311 | |||
L40374.1 | - | 4 | 586 |