Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003353.3 | NP_003344.1 | 2 | 573 | ||
GenBank mRNA | BC104470.1 | - | 2 | 374 | 40033510 | |
BC104471.2 | - | 2 | 374 | 40033515 | ||
CR542244.1 | - | 1 | 274 |