Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025366.2 | NP_001020537.2 | 1 | 1043 | ||
NM_001025367.2 | NP_001020538.2 | 1 | 1043 | |||
NM_001025368.2 | NP_001020539.2 | 1 | 1043 | |||
NM_001025369.2 | NP_001020540.2 | 1 | 1043 | |||
NM_001025370.2 | NP_001020541.2 | 1 | 1043 | |||
NM_001033756.2 | NP_001028928.1 | 1 | 1043 | |||
NM_001171622.1 | NP_001165093.1 | 1 | 1043 | |||
NM_001171623.1 | NP_001165094.1 | 1 | 1043 | |||
NM_001171624.1 | NP_001165095.1 | 1 | 1043 | |||
NM_001171625.1 | NP_001165096.1 | 1 | 1043 | |||
NM_001171626.1 | NP_001165097.1 | 1 | 1043 | |||
NM_001171627.1 | NP_001165098.1 | 1 | 1043 | |||
NM_001171628.1 | NP_001165099.1 | 1 | 1043 | |||
NM_001171629.1 | NP_001165100.1 | 1 | 1043 | |||
NM_001171630.1 | NP_001165101.1 | 1 | 1043 | |||
NM_001204384.1 | NP_001191313.1 | 1 | 1043 | |||
NM_001204385.1 | NP_001191314.1 | 1 | 1043 | |||
NM_001317010.1 | NP_001303939.1 | 1 | 1043 | |||
NM_003376.5 | NP_003367.4 | 1 | 1043 | |||
GenBank mRNA | AF022375.1 | - | 1 | 706 | ||
AY047581.1 | - | 1 | 21 | |||
BC011177.1 | - | 1 | 518 | |||
BC065522.1 | - | 1 | 470 | |||
BU153227.1 | - | 1 | 480 | |||
M27281.1 | - | 1 | 161 | |||
M32977.1 | - | 1 | 61 | |||
S85192.1 | - | 1 | 73 | |||
X62568.1 | - | 1 | 21 |