Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300951.1 | NP_001287880.1 | 3 | 1692 | ||
NM_003430.3 | NP_003421.2 | 4 | 1788 | |||
XM_006722880.3 | XP_006722943.1 | 2 | 1513 | |||
XM_011528265.2 | XP_011526567.1 | 4 | 1749 | |||
XM_011528266.2 | XP_011526568.1 | 4 | 1717 | |||
XM_017027235.1 | XP_016882724.1 | 4 | 1742 | |||
XM_017027236.1 | XP_016882725.1 | 3 | 1609 | |||
GenBank mRNA | AK291256.1 | - | 4 | 1695 | ||
AK300294.1 | - | 3 | 1578 | |||
L11672.1 | - | 4 | 1676 |