Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287054.1 | NP_001273983.1 | 4 | 679 | ||
NM_003439.2 | NP_003430.1 | 2 | 650 | |||
XM_005250565.4 | XP_005250622.1 | 2 | 641 | |||
XM_011516559.2 | XP_011514861.1 | 2 | 1205 | |||
GenBank mRNA | AF454940.1 | - | 2 | 621 | ||
AK054676.1 | - | 4 | 652 | |||
AK074116.1 | - | 2 | 608 | |||
AL833578.1 | - | 2 | 626 | |||
AY260738.1 | - | 2 | 623 | |||
BC022378.1 | - | 2 | 623 | 4685873 | ||
BC112224.1 | - | 1 | 453 | 8327692 | ||
BC113697.1 | - | 1 | 453 | 8322749 | ||
BX640646.1 | - | 2 | 623 |