Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173628.3 | NP_775899.3 | 58 | 9323 | ||
XM_011525416.2 | XP_011523718.1 | 58 | 9359 | |||
XM_017025261.1 | XP_016880750.1 | 16 | 2583 | |||
GenBank mRNA | AB290170.1 | - | 1 | 24 | ||
AK128065.1 | - | 5 | 803 | |||
AL832652.1 | - | 3 | 394 |