Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MATN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003833.4 | NP_003824.2 | 8 | 1317 | ||
NM_030590.3 | NP_085080.1 | 6 | 1136 | |||
NM_030592.3 | NP_085095.1 | 5 | 983 | |||
XM_005260597.1 | XP_005260654.1 | 7 | 1498 | |||
XM_017028113.1 | XP_016883602.1 | 8 | 1317 | |||
XM_017028114.1 | XP_016883603.1 | 6 | 1375 | |||
XM_017028115.1 | XP_016883604.1 | 5 | 1252 | |||
GenBank mRNA | AJ007581.1 | - | 6 | 1081 | ||
AK027323.1 | - | 2 | 173 | |||
AK074495.1 | - | 4 | 515 | |||
AK074593.1 | - | 9 | 1317 | |||
AK074595.1 | - | 6 | 1053 | |||
AK074597.1 | - | 6 | 1138 | |||
BC131763.1 | - | 6 | 1085 | 40121434 | ||
BC141811.1 | - | 6 | 1124 | 40147663 | ||
BC143625.1 | - | 6 | 1044 | |||
BC151219.1 | - | 7 | 1212 | 40148522 |