Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WISP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323369.1 | NP_001310298.1 | 3 | 1039 | ||
NM_001323370.1 | NP_001310299.1 | 5 | 1082 | |||
NM_003881.3 | NP_003872.1 | 4 | 1286 | |||
XM_017028116.1 | XP_016883605.1 | 3 | 597 | |||
GenBank mRNA | AF074604.1 | - | 4 | 745 | ||
AF083500.1 | - | 4 | 747 | |||
AF100780.1 | - | 4 | 894 | |||
AK129660.1 | - | 2 | 1543 | |||
AK313853.1 | - | 4 | 762 | |||
AY358915.1 | - | 4 | 748 | |||
BC017782.2 | - | 4 | 885 | 4691574 | ||
BC058074.1 | - | 3 | 988 | 5194313 | ||
BC064379.1 | - | 4 | 749 | 5725937 |