Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004184.3 | NP_004175.2 | 4, 5 | 813 | ||
NM_173701.1 | NP_776049.1 | 4, 5 | 589 | |||
NM_213645.1 | NP_998810.1 | 3, 4 | 417 | |||
NM_213646.1 | NP_998811.1 | 3, 4 | 641 | |||
XM_005268044.3 | XP_005268101.1 | 4, 5 | 541 | |||
XM_006720249.3 | XP_006720312.1 | 5, 6 | 775 | |||
XM_011537133.2 | XP_011535435.1 | 6, 7 | 787 | |||
XM_011537135.2 | XP_011535437.1 | 5, 6 | 671 | |||
XM_011537136.2 | XP_011535438.1 | 4, 5 | 507 | |||
XM_017021627.1 | XP_016877116.1 | 5, 6 | 679 | |||
XM_017021628.1 | XP_016877117.1 | 5, 6 | 810 | |||
XM_017021629.1 | XP_016877118.1 | 6, 7 | 785 | |||
GenBank mRNA | AK025487.1 | - | 2, 3 | 217 | ||
AK056100.1 | - | 4, 5 | 1051 | |||
AK291141.1 | - | 3, 4 | 393 | |||
AK300255.1 | - | 4, 5 | 696 | |||
BC017489.1 | - | 4, 5 | 689 | 3542671 | ||
BC095453.1 | - | 5, 6 | 700 | 30418785 | ||
BG717903.1 | - | 3, 4 | 422 | |||
BM561448.1 | - | 436 | ||||
BX248006.1 | - | 4, 5 | 533 | |||
M61715.1 | - | 4, 5 | 619 | |||
M77804.1 | - | 3, 4 | 474 | |||
X59892.1 | - | 4, 5 | 543 | |||
X62570.1 | - | 3, 4 | 494 |