Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOMER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145721.1 | NP_001139193.1 | 6 | 614 | ||
NM_001145722.1 | NP_001139194.1 | 6 | 1036 | |||
NM_001145724.1 | NP_001139196.1 | 4 | 474 | |||
NM_004838.3 | NP_004829.3 | 6 | 764 | |||
NR_027297.1 | 5 | 1168 | ||||
XM_006722943.1 | XP_006723006.1 | 6 | 648 | |||
XM_006722944.1 | XP_006723007.1 | 6 | 666 | |||
GenBank mRNA | AF093265.1 | - | 6 | 605 | ||
AK124450.1 | - | 5 | 1168 | |||
AK313995.1 | - | 6 | 1037 | |||
BC012113.1 | - | 6 | 706 | 4555641 | ||
BT019464.1 | - | 5 | 515 | |||
Y17573.1 | - | 5 | 515 | |||
Y18894.1 | - | 4 | 407 |