Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005.4 | NP_000996.2 | 3, 4 | 308 | ||
NM_001256802.1 | NP_001243731.1 | 3, 4 | 308 | |||
NM_001260507.1 | NP_001247436.1 | 2, 3 | 177 | |||
GenBank mRNA | AB062288.1 | - | 3, 4 | 244 | ||
AK129794.1 | - | 3, 4 | 280 | |||
AK130960.1 | - | 3, 4 | 281 | |||
AK223048.1 | - | 3, 4 | 257 | |||
AK313051.1 | - | 3, 4 | 277 | |||
BC003137.1 | - | 3, 4 | 257 | 2906158 | ||
BC003577.1 | - | 3, 4 | 249 | |||
BC013196.1 | - | 3, 4 | 249 | |||
BC013231.1 | - | 3, 4 | 269 | |||
BC029981.1 | - | 2, 3 | 202 | |||
BC034149.1 | - | 3, 4 | 281 | 4665438 | ||
BC071917.1 | - | 3, 4 | 260 | 6297012 | ||
BC100284.1 | - | 3, 4 | 269 | 6160514 | ||
BC150501.1 | - | 1, 2 | 83 | |||
BE797417.1 | - | 101 | ||||
BM831460.1 | - | 3, 4 | 307 | |||
S42658.1 | - | 3, 4 | 259 | |||
U14990.1 | - | 3, 4 | 251 | |||
U14991.1 | - | 3, 4 | 251 | |||
U14992.1 | - | 3, 4 | 251 | |||
X55715.1 | - | 3, 4 | 273 |