Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004965.6 | NP_004956.5 | 6 | 1108 | ||
XM_011529543.1 | XP_011527845.1 | 9 | 1835 | |||
XM_011529544.1 | XP_011527846.1 | 8 | 1789 | |||
XM_011529545.1 | XP_011527847.1 | 8 | 1169 | |||
XM_011529547.1 | XP_011527849.1 | 6 | 1018 | |||
XM_011529549.1 | XP_011527851.1 | 8 | 1330 | |||
XM_017028332.1 | XP_016883821.1 | 8 | 2619 | |||
XM_017028333.1 | XP_016883822.1 | 9 | 2691 | |||
XM_017028334.1 | XP_016883823.1 | 8 | 2601 | |||
XM_017028335.1 | XP_016883824.1 | 7 | 1123 | |||
XM_017028336.1 | XP_016883825.1 | 8 | 1252 | |||
GenBank mRNA | AB209245.1 | - | 5 | 4065 | ||
AK056033.1 | - | 3 | 1482 | |||
AK130775.1 | - | 2 | 689 | |||
BC000075.2 | - | 6 | 1032 | 3510312 | ||
BC023984.1 | - | 6 | 1024 | 4104558 | ||
BC070154.1 | - | 6 | 1030 | 6495327 | ||
BC071643.1 | - | 6 | 1020 | 5549431 | ||
BC072672.1 | - | 6 | 1021 | 6166317 | ||
BC106080.1 | - | 6 | 1041 | 3855939 | ||
BC107741.1 | - | 6 | 1118 | 6495117 | ||
J02621.1 | - | 6 | 1056 |