Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005345.5 | NP_005336.3 | 1 | 2374 | ||
GenBank mRNA | AB209818.1 | - | 1 | 2335 | ||
AK096017.1 | - | 2 | 1850 | |||
AK225692.1 | - | 1 | 2345 | |||
AK225699.1 | - | 1 | 2345 | |||
AK291295.1 | - | 1 | 2345 | |||
AK294182.1 | - | 2 | 2138 | |||
AK298056.1 | - | 2 | 2273 | |||
AK298098.1 | - | 2 | 1673 | |||
AK301574.1 | - | 2 | 2291 | |||
BC002453.2 | - | 1 | 2308 | 3345864 | ||
FJ224292.1 | - | 1 | 2621 |