Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005368.2 | NP_005359.1 | 3 | 944 | ||
NM_203377.1 | NP_976311.1 | 4 | 1036 | |||
NM_203378.1 | NP_976312.1 | 4 | 1019 | |||
XM_005261605.2 | XP_005261662.1 | 4 | 980 | |||
GenBank mRNA | AK300657.1 | - | 4 | 902 | ||
BC014547.1 | - | 3 | 940 | 4244838 | ||
BC018001.1 | - | 3 | 909 | 4271145 |